Tin(II) 2-ethylhexanoate, tech.

Supplier: Thermo Scientific Chemicals
Warning

18590-22 18590-36 18590-A3
CAAA18590-22EA 59.09 CAD
CAAA18590-22 CAAA18590-36 CAAA18590-A3
Tin(II) 2-ethylhexanoate, tech.
Tin (II) 2-ethylhexanoate

MDL: MFCD00002676


CAS No.: 301-10-0 Molecular Formula: C16H30O4Sn Formula Weight: 405.11 Flash Point: 110°(230°F) Physical Form: Liquid Density: 1.251 Refractive Index: 1.4940 Appearance: Colorless to pale yellow MDL No.: MFCD00002676

Formula: C₁₆H₃₀O₄Sn
Density: 1.251
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002676
CAS Number: 301-10-0
EINECS: 206-108-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR