Triammonium dodecamolybdophosphate hydrate, REacton®, Reagent Grade
Supplier: Thermo Scientific Chemicals
|
Warning
|
17984-22
17984-36
CAAA17984-22EA
135.09
CAD
CAAA17984-22
CAAA17984-36
Triammonium dodecamolybdophosphate hydrate, REacton®, Reagent Grade
Triammonium dodecamolybdophosphate hydrate
MDL: MFCD00036312
CAS No.: 54723-94-3
Molecular Formula: H12Mo12N3O40P.xH2O (x≈3)
Formula Weight: 1876.35(anhy)
Physical Form: Powder
Density: 3.15
Appearance: Yellow
Melting Point: dec.
MDL No.: MFCD00036312
Formula:
H₁₂Mo₁₂N₃O₄₀P·H₂O Melting Pt: Dec. Density: 3.15 Storage Temperature: Ambient |
MDL Number:
MFCD00150423 CAS Number: 54723-94-3 EINECS: 234-714-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...