Methyl orange ACS

Supplier: Thermo Scientific Chemicals
Danger

17874-06 17874-14 17874-22
CAAA17874-06EA 27.97 CAD
CAAA17874-06 CAAA17874-14 CAAA17874-22
Methyl orange ACS
Methyl orange

MDL: MFCD00007502 Beilstein Registry No.: 4732884 Soluble in 500 parts water. More soluble in hot water


CAS No.: 547-58-0 Molecular Formula: C14H14N3NaO3S Formula Weight: 327.34 Physical Form: Crystalline Appearance: Orange Odor: Weak, characteristic pH: 6.5 Melting Point: 300° Solubility: Soluble in 500 parts water. More soluble in hot water Beilstein Reference: 4732884 Merck Reference: 14,6105 UN No.: 3143 MDL No.: MFCD00007502

Formula: C₁₄H₁₄N₃NaO₃S
Boiling Pt: ∼90 °C (1013 hPa)
Melting Pt: >300 °C
Density: ∼1 g/cm³ (20 °C)
Flash Pt: 38 °C
Storage Temperature: Ambient
MDL Number: MFCD00007502
CAS Number: 547-58-0
EINECS: 208-925-3
UN: 3143
ADR: 6.1,III
Merck Index: 14,6105

Order Now

Specification Test Results

Clarity of solution Pass test
Visual transition interval pH 3.2(pink or red) to pH 4.4 (yellow)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR