Diammonium-L(+)-tartrate 98%
Supplier: Thermo Scientific Chemicals
17658-18
17658-30
17658-A1
CAAA17658-18EA
53.88
CAD
CAAA17658-18
CAAA17658-30
CAAA17658-A1
Diammonium-L(+)-tartrate 98%
di-Ammonium L(+)-tartrate
MDL: MFCD00013073
Beilstein Registry No.: 6120352
Soluble in water, alcohol
Purity: 98%
CAS No.: 3164-29-2
Molecular Formula: C4H12N2O6
Formula Weight: 184.15
Physical Form: Crystalline
Density: 1.6
Appearance: White
Odor: Odorless
Solubility: Soluble in water, alcohol
Beilstein Reference: 6120352
MDL No.: MFCD00013073
Formula:
C₄H₁₂N₂O₆ Density: 1.6 Storage Temperature: Ambient |
MDL Number:
MFCD00013073 CAS Number: 3164-29-2 EINECS: 221-618-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...