Bismuth triacetate 99%
Supplier: Thermo Scientific Chemicals
17574-18
17574-30
CAAA17574-18EA
213.3
CAD
CAAA17574-18
CAAA17574-30
Bismuth triacetate 99%
Bismuth triacetate
MDL: MFCD00015629
Purity: 99%
CAS No.: 22306-37-2
Molecular Formula: C6H9BiO6
Formula Weight: 386.12
Physical Form: Crystalline
Sensitivity: Hygroscopic
Appearance: White
Melting Point: dec.
MDL No.: MFCD00015629
Formula:
(H₃CCOO)₃Bi Melting Pt: Dec. Storage Temperature: Ambient |
MDL Number:
MFCD00015629 CAS Number: 22306-37-2 EINECS: 244-904-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...