Tetrapropylammonium hydroxide 40% (w/w) in aqueous solution

Supplier: Thermo Scientific Chemicals
Danger

17456-14 17456-22 17456-36
CAAA17456-06EA 54.98 CAD
CAAA17456-06 CAAA17456-14 CAAA17456-22 CAAA17456-36
Tetrapropylammonium hydroxide 40% (w/w) in aqueous solution
Tetrapropylammonium hydroxide

MDL: MFCD00009360 Beilstein Registry No.: 4207983


CAS No.: 4499-86-9 Molecular Formula: C12H29NO Formula Weight: 203.37 Physical Form: Liquid Density: 0.99 Refractive Index: 1.3716 Sensitivity: Air Sensitive Appearance: Colorless to pale yellow Boiling Point: 102° Beilstein Reference: 4207983 UN No.: 3267 MDL No.: MFCD00009360

Formula: C₁₂H₂₉NO
Boiling Pt: 102 °C
Density: 0.99
Storage Temperature: Ambient
MDL Number: MFCD00009360
CAS Number: 4499-86-9
EINECS: 224-800-6
UN: 3267
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR