Iron(II) sulfide ≥99% (metals basis)

Supplier: Thermo Scientific Chemicals

Synonyms: Ferrous sulphide, Ferrous sulfide

17422-09 17422-18
CAAA17422-09EA 96.54 CAD
CAAA17422-09 CAAA17422-18
Iron(II) sulfide ≥99% (metals basis)
Iron (II) sulphide

MDL: MFCD00011013 Soluble in acids with H2S evolution


Purity: 99% CAS No.: 1317-37-9 Molecular Formula: FeS (Fe typically 0.9) Formula Weight: 87.91 Physical Form: -100 Mesh Powder Density: 4.74 Appearance: Brown to black Odor: Odorless Melting Point: 1195° Solubility: Soluble in acids with H2S evolution Merck Reference: 14,4058 MDL No.: MFCD00011013

Formula: FeS
MW: 87.91 g/mol
Melting Pt: 1195
Density: 4.74
Storage Temperature: Ambient
MDL Number: MFCD00011013
CAS Number: 1317-37-9
EINECS: 215-268-6
UN: 3077
ADR: 9 III
Merck Index: 14,4058

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR