Arsenic (II) sulfide tech. 90%, Technical Grade
Supplier: Thermo Scientific Chemicals
|
Danger
|
17284-06
17284-14
CAAA17284-06EA
93.22
CAD
CAAA17284-06
CAAA17284-14
Arsenic (II) sulfide tech. 90%, Technical Grade
Arsenic (II) sulphide
MDL: MFCD00049841
Purity: 90%
CAS No.: 12044-79-0
Molecular Formula: As2S2
Formula Weight: 213.97
Physical Form: Powder
Density: 3.5
Melting Point: ≈307°
Boiling Point: 565°
UN No.: 1557
MDL No.: MFCD00049841
Formula:
As₂S₂ MW: 213.98 Boiling Pt: 565 Melting Pt: 360 Density: 3.5 Storage Temperature: Ambient |
MDL Number:
MFCD00049841 CAS Number: 1303-32-8 UN: 1557 ADR: 6.1 III Merck Index: 12,00834 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...