4,4-Dimethyl-2-cyclohexen-1-one 97+%

Supplier: Thermo Scientific Chemicals
Warning

17162-03 17162-06
CAAA17162-03EA 45.98 CAD
CAAA17162-03 CAAA17162-06
4,4-Dimethyl-2-cyclohexen-1-one 97+%
4,4-Dimethyl-2-cyclohexen-1-one

MDL: MFCD00009695


Purity: 97+% CAS No.: 1073-13-8 Molecular Formula: (CH3)2C6H6(=O) Formula Weight: 124.18 Flash Point: 64°(148°F) Physical Form: Liquid Density: 0.944 Sensitivity: Air Sensitive Boiling Point: 73-74°/14mm MDL No.: MFCD00009695

Formula: C₈H₁₂O
Boiling Pt: 73…74 °C (14 mmHg)
Density: 0,944
Flash Pt: 64 °C (148 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009695
CAS Number: 1073-13-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR