Bismuth(III) oxide ≥99.99% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Bismuth Trioxide, Dibismuth trioxide
17132-18
17132-30
CAAA17132-18EA
126.87
CAD
CAAA17132-18
CAAA17132-30
Bismuth(III) oxide ≥99.99% (metals basis)
Bismuth (III) oxide
MDL: MFCD00003462
Purity: 99.99%
CAS No.: 1304-76-3
Molecular Formula: Bi2O3
Formula Weight: 465.96
Physical Form: -325 Mesh Powder
Density: 8.9
Melting Point: 817°
Boiling Point: ca 1890°
Merck Reference: 14,1273
MDL No.: MFCD00003462
Formula:
BiO₃ MW: 465.96 g/mol Boiling Pt: ∼1890 °C Melting Pt: 817 °C Density: 8.9 Storage Temperature: Ambient |
MDL Number:
MFCD00003462 CAS Number: 1304-76-3 EINECS: 215-134-7 Merck Index: 14,1273 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...