Bismuth chloride oxide ≥99.95% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Bismuth oxychloride, Bismuth (III) oxychloride, Bismuth(III) chloride oxide
17131-14
17131-22
CAAA17131-14EA
134.14
CAD
CAAA17131-14
CAAA17131-22
Bismuth chloride oxide ≥99.95% (metals basis)
Bismuth chloride oxide
MDL: MFCD00010895
Purity: 99.95%
CAS No.: 7787-59-9
Molecular Formula: BiClO
Formula Weight: 260.43
Physical Form: Powder
Density: 7.72
Refractive Index: 2.15
Appearance: Very pale brown
Odor: Odorless
Melting Point: dec.
Merck Reference: 14,1263
MDL No.: MFCD00010895
Formula:
BiClO Melting Pt: Dec. Density: 7.72 Storage Temperature: Ambient |
MDL Number:
MFCD00010895 CAS Number: 7787-59-9 EINECS: 232-122-7 Merck Index: 14,1263 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...