Bismuth(III) bromide ≥99% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
17112-14
17112-22
CAAA17112-14EA
148.84
CAD
CAAA17112-14
CAAA17112-22
Bismuth(III) bromide ≥99% (metals basis)
Bismuth (III) bromide
MDL: MFCD00014195
Purity: 99%
CAS No.: 7787-58-8
Molecular Formula: BiBr3
Formula Weight: 448.71
Physical Form: Powder
Density: 5.72
Appearance: Yellow
Odor: Acidic
Melting Point: 218°
Boiling Point: 453°
UN No.: 3260
MDL No.: MFCD00014195
Formula:
BiBr₃ Boiling Pt: 453 °C Melting Pt: 218 °C Density: 5.72 Storage Temperature: Ambient |
MDL Number:
MFCD00014195 CAS Number: 7787-58-8 EINECS: 232-121-1 UN: 3260 ADR: 8,III Merck Index: 12,01299 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...