Amberlite® IRN-77, ion exchange resin cationic type (H⁺) nuclear grade

Supplier: Thermo Scientific Chemicals

16821-36 16821-A3
CAAA16821-36EA 74.1 CAD
CAAA16821-36 CAAA16821-A3
Amberlite® IRN-77, ion exchange resin cationic type (H⁺) nuclear grade
AMBERLITE® IRN-77, Ion exchange resin cationic type (H⁺)

MDL: MFCD00145822


CAS No.: 11128-94-2 Molecular Formula: Styrene-DVB; Strongly acidic cation exchange resin Merck Reference: 14,382 MDL No.: MFCD00145822

Storage Temperature: Ambient
MDL Number: MFCD00145822
CAS Number: 11128-94-2
Merck Index: 14,382

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR