Azomethine H hydrate
Supplier: Thermo Scientific Chemicals
Synonyms:
4-Hydroxy-5-[[(2-hydroxyphenyl)methylene]amino]naphthalene-2,7-disulphonic acid hydrate, 8-Hydroxy-1-[[(2-hydroxyphenyl)methylene]amino]naphthalene-3,6-disulphonic acid hydrate, 8-Hydroxy-1-(salicylideneamino)naphthalene-3,6-disulphonic acid hydrate, Azomethine-H hydrate, 4-Hydroxy-5-[[(2-hydroxyphenyl)methylene]amino]naphthalene-2,7-disulfonic acid hydrate, 8-Hydroxy-1-[[(2-hydroxyphenyl)methylene]amino]naphthalene-3,6-disulfonic acid hydrate, 8-Hydroxy-1-(salicylideneamino)naphthalene-3,6-disulfonic acid hydrate
MDL: MFCD00066536
Notes: Reagent for boron determination
CAS No.: 32266-60-7
Molecular Formula: C17H13NO8S2.xH2O
Formula Weight: 423.40(anhy)
Physical Form: Crystalline
MDL No.: MFCD00066536
Formula:
C₁₇H₁₃NO₈S₂·H₂O MW: 441.44 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00066536 CAS Number: 32266-60-7 EINECS: 250-975-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...