4-Bromo-1-butanol cont. varying amounts of THF, tech.
Supplier: Thermo Scientific Chemicals
|
Warning
|
16805-03
16805-06
16805-14
CAAA16805-03EA
106.65
CAD
CAAA16805-03
CAAA16805-06
CAAA16805-14
4-Bromo-1-butanol cont. varying amounts of THF, tech.
4-Bromo-1-butanol
MDL: MFCD00015387
CAS No.: 33036-62-3
Molecular Formula: C4H9BrO
Formula Weight: 153.02
Storage Temperature: -30°C to -10°C
Flash Point: 32°(91°F)
Physical Form: Liquid
Sensitivity: Moisture and Light Sensitive
Appearance: Yellow
Boiling Point: 63-65°/5mm
UN No.: 1987
MDL No.: MFCD00015387
Formula:
BrH₂C(CH₂)₂CH₂OH Boiling Pt: 63…65 °C (5 mmHg) Flash Pt: 32 °C (91 °F) Storage Temperature: Freezer |
MDL Number:
MFCD00015387 CAS Number: 33036-62-3 EINECS: 251-355-5 UN: 1987 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...