Trihexyl phosphate 90+%

Supplier: Thermo Scientific Chemicals
Warning

16801-06 16801-14 016801-03
CAAA16801-06EA 132.56 CAD
CAAA16801-06 CAAA16801-14 AA16801-03
Trihexyl phosphate 90+%
Trihexyl phosphate

MDL: MFCD00015351


Purity: 90+% CAS No.: 2528-39-4 Molecular Formula: C18H39O4P Formula Weight: 350.48 Physical Form: Liquid Appearance: Colorless MDL No.: MFCD00015351

Formula: C₁₈H₃₉O₄P
MW: 350.48 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00015351
CAS Number: 2528-39-4
EINECS: 219-774-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR