Barium bromide, anhydrous 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
16792-18
16792-30
CAAA16792-18EA
131.14
CAD
CAAA16792-18
CAAA16792-30
Barium bromide, anhydrous 99%
Barium bromide
MDL: MFCD00003444
Soluble in water
Purity: 99%
CAS No.: 10553-31-8
Molecular Formula: BaBr2
Formula Weight: 297.14
Physical Form: Powder
Density: 4.781
Appearance: White
Odor: Odorless
Melting Point: 850°
Solubility: Soluble in water
Merck Reference: 14,968
UN No.: 1564
MDL No.: MFCD00003444
Formula:
BaBr₂ Boiling Pt: 1835 °C (1013 hPa) Melting Pt: 850 °C Density: 4.781 Storage Temperature: Ambient |
MDL Number:
MFCD00003444 CAS Number: 10553-31-8 EINECS: 234-140-0 UN: 1564 ADR: 6.1,III Merck Index: 14,968 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...