Strontium bromide, anhydrous ≥99% (metals basis excluding Ba) (max. 0.1% Ba)

Supplier: Thermo Scientific Chemicals
Warning

16791-18 16791-30
CAAA16791-18EA 178.54 CAD
CAAA16791-18 CAAA16791-30
Strontium bromide, anhydrous ≥99% (metals basis excluding Ba) (max. 0.1% Ba)
Strontium bromide

MDL: MFCD00049548


Purity: 99% CAS No.: 10476-81-0 Molecular Formula: Br2Sr Formula Weight: 247.44 Physical Form: Granular Density: 4.216 Appearance: White Odor: Odorless Melting Point: 643° Merck Reference: 14,8837 MDL No.: MFCD00049548

Formula: Br₂Sr
MW: 247.43 g/mol
Boiling Pt: 2146 °C (1013 hPa)
Melting Pt: 643 °C
Density: 4.216
Storage Temperature: Ambient
MDL Number: MFCD00049548
CAS Number: 10476-81-0
EINECS: 233-969-5
Merck Index: 14,8837

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR