Strontium chloride, anhydrous ≥99.5% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

16790-14 16790-22 16790-36
CAAA16790-14EA 170.64 CAD
CAAA16790-14 CAAA16790-22 CAAA16790-36
Strontium chloride, anhydrous ≥99.5% (metals basis)
Strontium chloride

MDL: MFCD00011249


Purity: 99.5% CAS No.: 10476-85-4 Molecular Formula: Cl2Sr Formula Weight: 158.53 Physical Form: -40 Mesh Powder Density: 3.052 Refractive Index: 1.650 Sensitivity: Hygroscopic Appearance: White Odor: Odorless Melting Point: 873° Boiling Point: 1250° Merck Reference: 14,8840 MDL No.: MFCD00011249

Formula: Cl₂Sr
MW: 158.53
Boiling Pt: 1250 °C
Melting Pt: 873 °C
Density: 1.96
Storage Temperature: Ambient
MDL Number: MFCD00011249
CAS Number: 10476-85-4
EINECS: 233-971-6
Merck Index: 14,8840

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR