Diethyl ether, anhydrous 99 Stabilised ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Ethyl Ether, Ether, 2,2'-Oxydiethane, 2,2'-Oxybisethane, Ethoxyethane
Stabilization:
BHT
16767-18
16767-36
16767-62
CAAA16767-18EA
60.04
CAD
CAAA16767-18
CAAA16767-36
CAAA16767-62
Diethyl ether, anhydrous 99 Stabilised ACS
Diethyl ether
MDL: MFCD00011646
Notes: 0.7134, 1.35555
Purity: 99% min
CAS No.: 60-29-7
Molecular Formula: C4H10O
Formula Weight: 74.12
Flash Point: -45°(-49°F)
Physical Form: Liquid
Density: 0.713
Refractive Index: 1.3525
Appearance: Colorless
Odor: Sweet
Melting Point: -116°
Boiling Point: 34.6°
Merck Reference: 14,3806
UN No.: 1155
MDL No.: MFCD00011646
Formula:
(CH₃CH₂)₂O MW: 74.12 g/mol Boiling Pt: 35 Melting Pt: -123 Density: 0.713 Flash Pt: –45 °C (–49 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00011646 CAS Number: 60-29-7 EINECS: 200-467-2 UN: 1155 ADR: 3 I REACH: 01-2119535785-29 Merck Index: 14,3806 |
|
Specification Test Results
Color (APHA) | 10 |
Peroxide (as H2O2) | 1ppm ppm |
Evaporation residue | 0.001% |
Titratable acid | 0.0002meq/g |
Carbonyl (as HCHO) | 0.001% |
Alcohol (CH3CH2OH) | Pass test (limit about 0.05%) |
H2O | 0.03% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...