Hydrazine hydrate 99+%

Supplier: Thermo Scientific Chemicals
Danger

16651-57 16651-A3
CAAA16651-57EA 755.24 CAD
CAAA16651-57 CAAA16651-A3
Hydrazine hydrate 99+%
Hydrazine hydrate

Hydrazine Hydrate used as a reactant in the cyclizations of pyridinones. It is also used in the study of nanocrystal semiconductors, participating in the functionalization and passivation of surface states.


  • 64% H₂NNH₂ in H₂O


It is widely used as a reducing agent or an intermediate of synthesis in various industrial sectors like water treatment (effluents, industrial boilers), chemical treatment process (metals, mine extraction) or active ingredients synthesis (pharmaceuticals and agrochemicals).

Formula: NH₂NH₂·xH₂O
Boiling Pt: 120…121 °C
Melting Pt: –52 °C
Density: 1.027
Flash Pt: 75 °C (167 °F)
Storage Temperature: Ambient
MDL Number: MFCD00149931
CAS Number: 10217-52-4
EINECS: 206-114-9
UN: 2030
ADR: 8,II
Merck Index: 14,4771

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR