Cesium hydroxide 50% (w/w) in aqueous solution , 99% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

16505-MZ 16505-NF
CAAA16505-MZEA 1516.8 CAD
CAAA16505-MZ CAAA16505-NF
Cesium hydroxide 50% (w/w) in aqueous solution , 99% (metals basis)
Cesium hydroxide

MDL: MFCD00010964 Notes: Sold on Cesium hydroxide contained basis


Purity: 99% CAS No.: 21351-79-1 Molecular Formula: CsOH Formula Weight: 149.91 Physical Form: Liquid Density: 1.725 Appearance: Colorless Odor: Odorless UN No.: 2681 MDL No.: MFCD00010964

Formula: CsOH
Density: 1.725
Storage Temperature: Ambient
MDL Number: MFCD00010964
CAS Number: 21351-79-1
EINECS: 244-344-1
UN: 2681
ADR: 8,II
Merck Index: 13,02022

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR