Xylene (mixture of isomers) 98,5 isomers plus ethylbenzene ACS

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Dimethylbenzene

16371-AP 16371-K2 16371-K7 16371-M1
CAAA16371-APEA 64.27 CAD
CAAA16371-AP CAAA16371-K2 CAAA16371-K7 CAAA16371-M1
Xylene (mixture of isomers) 98,5 isomers plus ethylbenzene ACS
Xylene (mixture of isomers)

MDL: MFCD00077264 Beilstein Registry No.: 1901563 Notes: Contains a mixture of the ortho, meta, and para isomers of xylene. May also contain ethylbenzene.


Purity: 98.5+% CAS No.: 1330-20-7 Molecular Formula: (CH3)2C6H4 Formula Weight: 106.17 Flash Point: 29°(84°F) Physical Form: Liquid Density: 0.860 Refractive Index: 1.4970 Odor: Aromatic Melting Point: -25° Boiling Point: 137-144° Beilstein Reference: 1901563 Merck Reference: 14,10081 UN No.: 1307 MDL No.: MFCD00077264

Formula: C₆H₄(CH₃)₂
MW: 106.17 g/mol
Boiling Pt: 140
Melting Pt: < –25 °C
Density: 0.860
Flash Pt: 29 °C (84 °F)
Storage Temperature: Ambient
MDL Number: MFCD00077264
CAS Number: 1330-20-7
EINECS: 215-535-7
UN: 1307
ADR: 3 III
Merck Index: 14,10081
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2deefc7b7b67c10e64003e243ff3216fad27cf7d&vid=9fa01ddafd4814d5f34266ae7933463a29d92d19&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Color (APHA) 10
Evaporation residue 0.002 max.
Substances darkened by sulfuric acid Pass Test
Sulfur compounds (as S) 0.003% max.
H2O 0.05% max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR