Tetraethylammonium hydroxide 25% (w/w) in methanol
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Tetrylammonium hydroxide
16328-18
16328-30
CAAA16328-18EA
221.2
CAD
CAAA16328-18
CAAA16328-30
Tetraethylammonium hydroxide 25% (w/w) in methanol
Tetraethylammonium hydroxide
MDL: MFCD00009024
Beilstein Registry No.: 3912961
CAS No.: 77-98-5
Molecular Formula: (CH3CH2)4NOH
Formula Weight: 147.26
Flash Point: 16°(60°F)
Physical Form: Liquid
Density: 0.88
Refractive Index: 1.380
Sensitivity: Air Sensitive
Odor: Like alcohol
Boiling Point: dec.
Beilstein Reference: 3912961
Merck Reference: 14,9201
UN No.: 2924
MDL No.: MFCD00009024
Formula:
C₈H₂₁NO MW: 147.26 g/mol Boiling Pt: Dec. Density: 0.88 Flash Pt: 16 °C (60 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009024 CAS Number: 77-98-5 EINECS: 201-073-3 UN: 2924 ADR: 3,II Merck Index: 14,9201 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...