Pyrazine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
MDL: MFCD00006122
Beilstein Registry No.: 103905
Soluble in water, alcohol, and ether
Purity: 98%
CAS No.: 290-37-9
Molecular Formula: C4H4N2
Formula Weight: 80.09
Flash Point: 55°(131°F)
Physical Form: Crystalline
Density: 1.031
Refractive Index: 1.5235
Sensitivity: Hygroscopic
Appearance: White
Odor: Strong
Melting Point: 50-56°
Boiling Point: 113-116°
Solubility: Soluble in water, alcohol, and ether
Beilstein Reference: 103905
Merck Reference: 14,7957
UN No.: 1325
MDL No.: MFCD00006122
Formula:
C₄H₄N₂ Boiling Pt: 113…116 °C Melting Pt: 50…56 °C Density: 1.031 Flash Pt: 55 °C (131 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006122 CAS Number: 290-37-9 EINECS: 206-027-6 UN: 1325 ADR: 4.1,III Merck Index: 14,7957 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...