Thymol blue ACS
Supplier: Thermo Scientific Chemicals
16272-04
16272-09
16272-18
CAAA16272-04EA
38.71
CAD
CAAA16272-04
CAAA16272-09
CAAA16272-18
Thymol blue ACS
Thymol blue
pH indicator: 1.2 (red) to 2.8 (yellow) to 9.6 (blue)
CAS No.: 76-61-9
Molecular Formula: C27H30O5S
Formula Weight: 466.60
Physical Form: Crystalline
Melting Point: 223° dec.
Solubility: Insoluble in water. Soluble in alcohol, dilute alkali solutions
MDL No.: MFCD00005869
pH indicator: 1.2 (red) to 2.8 (yellow) to 9.6 (blue)
Formula:
C₂₇H₃₀O₅S Melting Pt: 223 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00005869 CAS Number: 76-61-9 EINECS: 200-973-3 Merck Index: 14,9400 |
Specification Test Results
Clarity of solution | Pass test |
Visual transition interval (acid range) | pH 1.2 (red) to 2.8 (yellow) |
Visual transition range (alkaline range) | pH 8.0 (yellow) to 9.2 (blue) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...