Tetrabutylammonium tribromide 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
16258-14
16258-22
CAAA16258-14EA
89.43
CAD
CAAA16258-14
CAAA16258-22
Tetrabutylammonium tribromide 98%
Tetrabutylammonium tribromide
MDL: MFCD00012110
Beilstein Registry No.: 3746114
Purity: 98%
CAS No.: 38932-80-8
Molecular Formula: C16H36Br3N
Formula Weight: 482.18
Physical Form: Crystalline
Appearance: Yellow
Melting Point: 74-76°
Beilstein Reference: 3746114
UN No.: 3261
MDL No.: MFCD00012110
Formula:
C₁₆H₃₆Br₃N Melting Pt: 74…76 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012110 CAS Number: 38932-80-8 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...