Tetraethylammonium hexafluorophosphate 98%

Supplier: Thermo Scientific Chemicals
Danger

16256-09 16256-18
CAAA16256-09EA 127.19 CAD
CAAA16256-09 CAAA16256-18
Tetraethylammonium hexafluorophosphate 98%
Tetraethylammonium hexafluorophosphate

MDL: MFCD00043179


Purity: 98% CAS No.: 429-07-2 Molecular Formula: (C2H5)4NPF6 Formula Weight: 275.23 Physical Form: Crystalline Sensitivity: Hygroscopic UN No.: 2811 MDL No.: MFCD00043179

Formula: C₈H₂₀F₆NP
Storage Temperature: Ambient
MDL Number: MFCD00043179
CAS Number: 429-07-2
EINECS: 207-056-7
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR