Phenol red sodium salt ACS
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3,3-Bis(4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide, sodium salt, Phenol red water soluble, Sodium hydrogen 4,4'-(3H-2,1-benzoxathiol-3-ylidene)bisphenolate S,S-dioxide
15987-06
15987-14
CAAA15987-06EA
51.35
CAD
CAAA15987-06
CAAA15987-14
Phenol red sodium salt ACS
Phenol red sodium salt
pH indicator
CAS No.: 34487-61-1
Molecular Formula: C19H13NaO5S
Formula Weight: 376.37
Physical Form: Crystalline
Melting Point: 285° dec.
Beilstein Reference: 6260026
Merck Reference: 14,7247
MDL No.: MFCD00066901
pH indicator
Formula:
C₁₉H₁₃NaO₅S MW: 376.35 g/mol Melting Pt: 285 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00066901 CAS Number: 34487-61-1 EINECS: 252-057-8 Merck Index: 14,7247 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...