Benzo[a]pyrene ≥96%
Supplier: Thermo Scientific Chemicals
|
Danger
|
15856-02
15856-MC
CAAA15856-02EA
401.32
CAD
CAAA15856-02
CAAA15856-MC
Benzo[a]pyrene ≥96%
Benzo[a]pyrene
Extensively used in cancer research
Purity: 96%
CAS No.: 50-32-8
Molecular Formula: C20H12
Formula Weight: 252.30
Physical Form: Crystalline
Appearance: Pale yellow/green/orange
Melting Point: 177-180°
Boiling Point: 495°
Solubility: Soluble in benzene, toluene, and xylene. Sparingly soluble in alcohol, methanol. Insoluble in water
Merck Reference: 14,1103
UN No.: 3077
MDL No.: MFCD00003602
Extensively used in cancer research
Formula:
C₂₀H₁₂ Boiling Pt: 495 °C Melting Pt: 177…180 °C Density: 1.351 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003602 CAS Number: 50-32-8 EINECS: 200-028-5 UN: 3077 ADR: 9,III Merck Index: 14,1103 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...