Perfluorohexane 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Tetradecafluorohexane

15812-36 15812-14 15812-22
CAAAA15812-36EA 521.4 CAD
CAAAA15812-36 CAAA15812-14 CAAA15812-22
Perfluorohexane 98+%
Perfluorohexane

MDL: MFCD00000437 Beilstein Registry No.: 1802113 Notes: 85% n-isomer


Purity: 98+% CAS No.: 355-42-0 Molecular Formula: C6F14 Formula Weight: 338.04 Flash Point: None Physical Form: Clear colorless liquid Density: 1.699 Refractive Index: 1.2510 Appearance: Colorless Melting Point: -4° Boiling Point: 58-60° Beilstein Reference: 1802113 MDL No.: MFCD00000437

Formula: C₆F₁₄
Boiling Pt: 58…60 °C
Melting Pt: –4 °C
Density: 1,699
Storage Temperature: Ambient
MDL Number: MFCD00000437
CAS Number: 355-42-0
EINECS: 206-585-0

Order Now

Specification Test Results

Form Clear colorless liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR