Oxalacetic acid 98+%

Supplier: Thermo Scientific Chemicals
Danger

15789-04 15789-09
CAAA15789-04EA 68.73 CAD
CAAA15789-04 CAAA15789-09
Oxalacetic acid 98+%
Oxalacetic acid

MDL: MFCD00002592 Beilstein Registry No.: 1705475


Purity: 98+% CAS No.: 328-42-7 Molecular Formula: C4H4O5 Formula Weight: 132.07 Flash Point: 88°(190°F) Physical Form: Powder Melting Point: 161° dec. Beilstein Reference: 1705475 Merck Reference: 14,6909 UN No.: 3261 MDL No.: MFCD00002592

Formula: C₄H₄O₅
Boiling Pt: 310 °C (1013 hPa)
Melting Pt: 161 °C (dec.)
Flash Pt: 88 °C (190 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002592
CAS Number: 328-42-7
EINECS: 206-329-8
UN: 3261
ADR: 8,II
Merck Index: 14,6909

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR