1-Octanesulfonic acid sodium salt monohydrate 99+%

Supplier: Thermo Scientific Chemicals

Synonyms: Sodium 1-octansulphonate monohydrate, Sodium 1-octansulfonate monohydrate, Sodium 1-octanesulphonate monohydrate

15764-09 15764-18
CAAA15764-09EA 125.61 CAD
CAAA15764-09 CAAA15764-18
1-Octanesulfonic acid sodium salt monohydrate 99+%
1-Octanesulphonic acid sodium salt monohydrate

MDL: MFCD00007544


Purity: 99+% CAS No.: 207596-29-0 Molecular Formula: CH3(CH2)7SO3Na.H2O Formula Weight: 234.29 (216.28anhy) Physical Form: Crystalline Melting Point: 300° MDL No.: MFCD00007544

Formula: C₈H₁₇NaO₃S·1H₂O
MW: 234.29 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00007544
CAS Number: 207596-29-0
EINECS: 226-195-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR