1-Pentanesulfonic acid sodium salt monohydrate for HPLC

Supplier: Thermo Scientific Chemicals

Synonyms: Sodium 1-pentanesulfonate monohydrate, Sodium 1-pentanesulphonate monohydrate

15714-06 15714-14 15714-22
CAAA15714-06EA 86.58 CAD
CAAA15714-06 CAAA15714-14 CAAA15714-22
1-Pentanesulfonic acid sodium salt monohydrate for HPLC
1-Pentanesulphonic acid sodium salt monohydrate

Ion pair reagent


CAS No.: 207605-40-1 Molecular Formula: C5H11NaO3S.H2O Formula Weight: 192.21 (174.19anhy) Physical Form: Crystalline powder Appearance: White Beilstein Reference: 3725147


Ion pair reagent

Formula: C₅H₁₁NaO₃S·1H₂O
MW: 192.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00007541
CAS Number: 207605-40-1
EINECS: 245-208-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR