Methacrylic acid 99+% stabilised
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2-Methylpropenoic acid
Stabilization:
Stabilised with hydroquinone monomethyl ether (4-methoxyphenol, MEHQ) 250 ppm
MDL: MFCD00002651
Beilstein Registry No.: 1719937
Purity: 99+%
CAS No.: 79-41-4
Molecular Formula: CH2=C(CH3)CO2H
Formula Weight: 86.09
Flash Point: 65°(149°F)
Physical Form: Liquid
Density: 1.015
Refractive Index: 1.4310
Odor: Acrid
pH: 2.0-2.2
Melting Point: 16°
Boiling Point: 163°
Beilstein Reference: 1719937
Merck Reference: 14,5941
UN No.: 2531
MDL No.: MFCD00002651
Formula:
C₄H₆O₂ Boiling Pt: 163 °C Melting Pt: 16 °C Density: 1.015 Flash Pt: 65 °C (149 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002651 CAS Number: 79-41-4 EINECS: 201-204-4 UN: 2531 ADR: 8,II Merck Index: 14,5941 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...