Ethyl propionate
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Propionic acid ethyl ester
15498-A1
15498-D5
CAAA15498-A1EA
99.22
CAD
CAAA15498-A1
CAAA15498-D5
Ethyl propionate
Ethyl propionate
MDL: MFCD00009308
Beilstein Registry No.: 506287
Fieser: 1,385
CAS No.: 105-37-3
Molecular Formula: CH3CH2CO2CH2CH3
Formula Weight: 102.13
Flash Point: 12°(54°F)
Physical Form: Liquid
Density: 0.891
Refractive Index: 1.3850
Melting Point: -73°
Boiling Point: 99°
Beilstein Reference: 506287
Merck Reference: 14,3847
UN No.: 1195
MDL No.: MFCD00009308
Formula:
C₅H₁₀O₂ MW: 102.13 g/mol Boiling Pt: 99 °C Melting Pt: –73 °C Density: 0.891 Flash Pt: 12 °C (54 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009308 CAS Number: 105-37-3 EINECS: 203-291-4 UN: 1195 ADR: 3 II Merck Index: 14,3847 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...