Cerium triacetate sesquihydrate ≥99.9% (REO, rare earth oxide basis)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Cerium (III) acetate sesquihydrate, Acetic acid cerium (III) salt sesquihydrate

15294-22 15294-36
CAAA15294-22EA 170.64 CAD
CAAA15294-22 CAAA15294-36
Cerium triacetate sesquihydrate ≥99.9% (REO, rare earth oxide basis)
Cerium triacetate sesquihydrate

With bromide ion, catalyzes the liquid-phase auto-oxidation of cresols.


Cerium(III) acetate sesquihydrate, acts as a source of cerium involved in the preparation of cerium oxide. It finds application in metallurgy, glass and glass polishing, ceramics, catalysts, and in phosphors. It is utilized in steel production to remove the free sulfur and oxygen. Further, it is an efficient glass polishing agent.


With bromide ion, catalyzes the liquid-phase auto-oxidation of cresols

Formula: Ce(C₂H₃O₂)₃·1.5H₂O
MW: 1272.24
Melting Pt: 308 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00051060
CAS Number: 537-00-8
EINECS: 208-654-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR