1-Heptanesulfonic acid sodium salt monohydrate 99%

Supplier: Thermo Scientific Chemicals

Synonyms: Sodium 1-heptanesulfonate monohydrate, Sodium 1-heptanesulphonate monohydrate

15214-09 15214-18
CAAA15214-09EA 107.76 CAD
CAAA15214-09 CAAA15214-18
1-Heptanesulfonic acid sodium salt monohydrate 99%
1-Heptanesulphonic acid sodium salt monohydrate

MDL: MFCD00149550 Beilstein Registry No.: 3731762


Purity: 99% CAS No.: 207300-90-1 Molecular Formula: C7H15NaO3S.H2O Formula Weight: 220.26 (202.24anhy) Physical Form: Crystalline Appearance: White Melting Point: 300° Beilstein Reference: 3731762 MDL No.: MFCD00149550

Formula: C₇H₁₅NaO₃S·1H₂O
MW: 220.27 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00149550
CAS Number: 207300-90-1
EINECS: 245-210-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR