Barium iodide hydrate ≥95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
15126-14
15126-22
CAAA15126-14EA
136.35
CAD
CAAA15126-14
CAAA15126-22
Barium iodide hydrate ≥95%
Barium iodide hydrate
MDL: MFCD00003452
Freely soluble in water. Soluble in alcohol and acetone
Purity: 95%
CAS No.: 13718-50-8
Molecular Formula: BaI2.xH2O
Formula Weight: 391.15(anhy)
Physical Form: Powder
Sensitivity: Light Sensitive and Hygroscopic
Appearance: White
Odor: Odorless
Solubility: Freely soluble in water. Soluble in alcohol and acetone
Merck Reference: 14,980
UN No.: 1564
MDL No.: MFCD00003452
Formula:
BaI₂·H₂O Density: 5.15 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149155 CAS Number: 7787-33-9 EINECS: 237-276-9 UN: 1564 ADR: 6.1,III Merck Index: 14,980 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...