Ethanesulfonic acid ≥96%

Supplier: Thermo Scientific Chemicals
Danger

14891-09 14891-18 14891-30
CAAA14891-09EA 79.79 CAD
CAAA14891-09 CAAA14891-18 CAAA14891-30
Ethanesulfonic acid ≥96%
Ethanesulphonic acid

MDL: MFCD00007529 Beilstein Registry No.: 1743071


Purity: 96% CAS No.: 594-45-6 Molecular Formula: CH3CH2SO3H Formula Weight: 110.13 Flash Point: 110°(230°F) Physical Form: Liquid Density: 1.343 Refractive Index: 1.4350 Appearance: Colorless to yellow Melting Point: -17° Boiling Point: 122-123°/0.01mm Beilstein Reference: 1743071 UN No.: 2586 MDL No.: MFCD00007529

Formula: C₂H₆O₃S
MW: 110.13 g/mol
Boiling Pt: 122…123 °C (0.01 mmHg)
Melting Pt: –17 °C
Density: 1,343
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007529
CAS Number: 594-45-6
EINECS: 209-843-0
UN: 2586
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR