Formamide ≥99.5% ACS

Supplier: Thermo Scientific Chemicals
Danger

14835-30 14835-A1 14835-D6
CAAA14835-30EA 118.18 CAD
CAAA14835-30 CAAA14835-A1 CAAA14835-D6
Formamide ≥99.5% ACS
Formamide

MDL: MFCD00007941


Purity: 99.5+% CAS No.: 75-12-7 Molecular Formula: CH3NO Formula Weight: 45.04 Flash Point: 154°(310°F) Physical Form: Liquid Density: 1.134 Refractive Index: 1.4475 Sensitivity: Hygroscopic Appearance: Colorless Odor: Ammonia-like Melting Point: 2° Boiling Point: 210° Beilstein Reference: 505995 Merck Reference: 14,4237 MDL No.: MFCD00007941

Formula: HCONH₂
Boiling Pt: 210 °C
Melting Pt: 2 °C
Density: 1.134
Flash Pt: 154 °C (310 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007941
CAS Number: 75-12-7
EINECS: 200-842-0
Merck Index: 14,4237
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f381691a96baf5054ca8291e1fa9e9603b687e31&vid=e54dda1a985d7a4f9485c2cc5233498087e43f88&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Color (APHA) 10
Freezing point 2.0-3.0°


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR