Silver ≥99.9% (metals basis), flakes 80% <20 µm

Supplier: Thermo Scientific Chemicals

14755-09 14755-50
CAAA14755-09EA 129.56 CAD
CAAA14755-09 CAAA14755-50
Silver ≥99.9% (metals basis), flakes 80% <20 µm
Silver

Suitable for manufacturing metallizing preparations and conductive silicone rubbers


Purity: 99.9% CAS No.: 7440-22-4 Odor: Characteristic UN No.: 3077 MDL No.: MFCD00003397


Suitable for manufacturing metallizing preparations and conductive silicone rubbers

Formula: Ag
Boiling Pt: 2212 °C (1013 hPa)
Melting Pt: 962 °C
Density: 10.5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003397
CAS Number: 7440-22-4
EINECS: 231-131-3
UN: 3077
ADR: 9,III
Merck Index: 12,08647

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR