Gold ≥99.95% (metals basis), wire, Ø 0.5 mm (0.02 in)
Supplier: Thermo Scientific Chemicals
14728-BS
14728-BW
14728-CB
CAAA14728-BSEA
195.92
CAD
CAAA14728-BS
CAAA14728-BW
CAAA14728-CB
Gold ≥99.95% (metals basis), wire, Ø 0.5 mm (0.02 in)
Gold
MDL: MFCD00003436
Purity: 99.95%
CAS No.: 7440-57-5
Physical Form: ≈3.79g/m
Odor: Odorless
MDL No.: MFCD00003436
Formula:
Au Boiling Pt: 2800 °C (1013 hPa) Melting Pt: 1063 °C Density: 19.3 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003436 CAS Number: 7440-57-5 EINECS: 231-165-9 Merck Index: 12,04526 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...