Gold ≥99.999% (metals basis), wire, Premion®, Ø 0.5 mm (0.02 in)

Supplier: Thermo Scientific Chemicals

14725-BS 14725-BW 14725-CB
CAAA14725-BSEA 238.58 CAD
CAAA14725-BS CAAA14725-BW CAAA14725-CB
Gold ≥99.999% (metals basis), wire, Premion®, Ø 0.5 mm (0.02 in)
Gold

MDL: MFCD00003436


Purity: 99.999% CAS No.: 7440-57-5 Physical Form: ≈3.79g/m Odor: Odorless MDL No.: MFCD00003436

Formula: Au
Boiling Pt: 2800 °C (1013 hPa)
Melting Pt: 1063 °C
Density: 19.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9
Merck Index: 12,04526

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR