Titanium(IV) chloride ≥99.0% (typ. 99.6%)
Supplier: Thermo Scientific Chemicals
|
Danger
|
In manufacture of titanium pigments and metal, as a catalyst in organic syntheses, in iridescent glass, as a smoke screen
Purity: 99.0% min
CAS No.: 7550-45-0
Molecular Formula: Cl4Ti
Formula Weight: 189.69
Physical Form: Liquid
Density: 1.726
Refractive Index: 1.61
Sensitivity: Moisture Sensitive
Appearance: Colorless
Melting Point: -25°
Boiling Point: 136-137°
Solubility: Soluble in dilute hydrochloric acid, alcohol and alkalies.
Merck Reference: 14,9478
UN No.: 1838
MDL No.: MFCD00011267
In manufacture of titanium pigments and metal, as a catalyst in organic syntheses, in iridescent glass, as a smoke screen
Formula:
Cl₄Ti Boiling Pt: 136…137 °C Melting Pt: –25 °C Density: 1.726 Storage Temperature: Ambient |
MDL Number:
MFCD00011267 CAS Number: 7550-45-0 EINECS: 231-441-9 UN: 1838 ADR: 6.1,I Merck Index: 14,9478 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...