Tantalum(V) oxide ≥99% (metals basis)
Supplier: Thermo Scientific Chemicals
Synonyms:
Ditantalum pentaoxide
14709-09
14709-18
14709-30
CAAA14709-09EA
71.42
CAD
CAAA14709-09
CAAA14709-18
CAAA14709-30
Tantalum(V) oxide ≥99% (metals basis)
Tantalum (V) oxide
MDL: MFCD00011254
Insoluble in water, alcohol, mineral acids. Soluble in HF
Purity: 99%
CAS No.: 1314-61-0
Molecular Formula: Ta2O5
Formula Weight: 441.89
Physical Form: Powder
Density: 8.2
Melting Point: 1800°
Solubility: Insoluble in water, alcohol, mineral acids. Soluble in HF
Merck Reference: 14,9056
MDL No.: MFCD00011254
Formula:
O₅Ta₂ MW: 441.89 g/mol Melting Pt: 1800 °C Density: 8.2 Storage Temperature: Ambient |
MDL Number:
MFCD00011254 CAS Number: 1314-61-0 EINECS: 215-238-2 Merck Index: 14,09056 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...