Tantalum pentachloride ≥99.99% (metals basis), Puratronic®

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Tantalum (V) chloride

14708-06 14708-14 14708-22
CAAA14708-06EA 86.43 CAD
CAAA14708-06 CAAA14708-14 CAAA14708-22
Tantalum pentachloride ≥99.99% (metals basis), Puratronic®
Tantalum pentachloride

MDL: MFCD00011253 Fieser: 16,312 17,321 18,343 19,318 20,355 21,407


Purity: 99.99% CAS No.: 7721-01-9 Molecular Formula: TaCl5 Formula Weight: 358.21 Physical Form: -60 Mesh Resublimed Powder, Ampouled under argon Density: 3.68 Appearance: White to pale yellow Odor: Acidic Melting Point: 216° Boiling Point: 242° Merck Reference: 14,9054 UN No.: 3260 MDL No.: MFCD00011253

Formula: Cl₅Ta
MW: 358.21 g/mol
Boiling Pt: 242 °C
Melting Pt: 216 °C
Density: 3.68
Storage Temperature: Ambient
MDL Number: MFCD00011253
CAS Number: 7721-01-9
EINECS: 231-755-6
UN: 3260
ADR: 8 III
Merck Index: 14,9054

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR