Sodium hexametaphosphate 66.5% P2O5 min., tech.

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Sodium metaphosphateSHMP

14706-A7 14706-A7 14706-A1 14706-A1
CAAA14706-A1EA 99.38 CAD
AA14706-A7 AA14706-A1 CAAA14706-A1 CAAA14706-A7
Sodium hexametaphosphate 66.5% P2O5 min., tech.
Sodium hexametaphosphate
Formula: Na₆P₆O₁₈
MW: 611.77 g/mol
Boiling Pt: 1500 °C
Melting Pt: 628 °C
Density: 1.25 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00136045
CAS Number: 10124-56-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR