Nickel(II) chloride, anhydrous ≥99% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Nickel dichloride

14687-22 14687-36
CAAA14687-22EA 199.08 CAD
CAAA14687-22 CAAA14687-36
Nickel(II) chloride, anhydrous ≥99% (metals basis)
Nickel (II) chloride

MDL: MFCD00011142 Fieser: 12,336 18,249 20,258 21,281


Purity: 99% CAS No.: 7718-54-9 Molecular Formula: Cl2Ni Formula Weight: 129.60 Physical Form: Powder Density: 3.55 Appearance: Orange Odor: Odorless Melting Point: 1001° Merck Reference: 14,6505 UN No.: 3288 MDL No.: MFCD00011142

Formula: Cl₂Ni
MW: 129.6 g/mol
Melting Pt: 1001 °C
Density: 3.55
Storage Temperature: Ambient
MDL Number: MFCD00011142
CAS Number: 7718-54-9
EINECS: 231-743-0
UN: 3288
ADR: 6.1 III
Merck Index: 14,6505

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR