Barium chromate 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
14669-30
14669-A1
CAAA14669-30EA
130.03
CAD
CAAA14669-30
CAAA14669-A1
Barium chromate 98%
Barium chromate
MDL: MFCD00014180
Insoluble in water, acetic acid, chromic acid
Purity: 98%
CAS No.: 10294-40-3
Molecular Formula: BaCrO4
Formula Weight: 253.33
Physical Form: Powder
Density: 4.50
Appearance: Yellow
Odor: Odorless
Solubility: Insoluble in water, acetic acid, chromic acid
Merck Reference: 14,972
UN No.: 1479
MDL No.: MFCD00014180
Formula:
BaCrO₄ Melting Pt: 1400 °C Density: 4.50 Storage Temperature: Ambient |
MDL Number:
MFCD00014180 CAS Number: 10294-40-3 EINECS: 233-660-5 UN: 1479 ADR: 5.1,III Merck Index: 14,972 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...